siRNA Details
Gene Symbol: LOC283911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833480.1 | - | 1 | 443 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL704195.1 | - | 1 | 441 | Chr. 16 - 35148637 - 35149311 [+] on Build GRCh38 |
