siRNA Details
Gene Symbol: LOC101928944 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120570.1 | 1 | 116 | |||
GenBank mRNA | AY726583.1 | - | 1 | 116 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI520577.1 | - | 94 | Chr. - - [-] on Build |
