siRNA Details
Gene Symbol: LINC00222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033376.1 | 4 | 713 | |||
GenBank mRNA | AY927447.1 | - | LIT3475 AY927447 |
4 | 676 | |
AY927448.1 | - | LIT3476 AY927448 |
4 | 555 |
