siRNA Details
Gene Symbol: FBXO31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282683.1 | NP_001269612.1 | 10 | 4988 | ||
NM_024735.4 | NP_079011.3 | 9 | 5240 | |||
GenBank mRNA | BX106211.1 | - | 1 | 133 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR621785.1 | - | 1 | 1672 | Chr. 16 - 87327016 - 87329388 [-] on Build GRCh38 |
