siRNA Details
Gene Symbol: ZFP69B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005271137.2 | XP_005271194.1 | 6 | 3916 | ||
XM_005271139.3 | XP_005271196.1 | 5 | 3864 | |||
GenBank mRNA | AK094134.1 | - | 1 | 1202 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA801448.1 | - | 1 | 146 | Chr. 1 - 40465375 - 40465946 [+] on Build GRCh38 |
