siRNA Details
Gene Symbol: LINC01305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC043589.1 | - | 2 | 100 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI913692.1 | - | 2 | 99 | Chr. 2 - 174328783 - 174329726 [+] on Build GRCh38 |
