siRNA Details
Gene Symbol: LOC101928797 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109806.1 | 2 | 131 | |||
GenBank mRNA | BC034724.1 | - | 2 | 131 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE893170.1 | - | 2 | 111 | Chr. 9 - 117648626 - 117656524 [+] on Build GRCh38 |
