siRNA Details
Gene Symbol: ANKRD20A19P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073430.1 | 3 | 3839 | |||
GenBank mRNA | AK127292.1 | - | 3 | 3839 | ||
AK128225.1 | - | 3 | 1306 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI833282.1 | - | 4 | 563 | Chr. 13 - 23940969 - 23946224 [-] on Build GRCh38 |
