siRNA Details
Gene Symbol: LOC645769 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123791.1 | - | 1 | 629 |
Gene Symbol: LOC107985657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030004.1 | XP_016885493.1 | 1 | 2079 |
