siRNA Details
Gene Symbol: HOXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524707.2 | XP_011523009.1 | 1 | 297 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA746255.1 | - | 1 | 298 | Chr. 17 - 48574311 - 48574798 [-] on Build GRCh38 |
