siRNA Details
Gene Symbol: LOC440934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136638.1 | 2 | 1061 | |||
NR_136639.1 | 2 | 765 | ||||
NR_136640.1 | 2 | 330 | ||||
NR_136641.1 | 2 | 509 | ||||
GenBank mRNA | AA401699.1 | - | 2 | 316 | ||
BC008048.2 | - | 2 | 1060 | |||
BC033986.1 | - | 2 | 731 |
