siRNA Details
Gene Symbol: LOC440934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136638.1 | 1 | 254 | |||
NR_136642.1 | 1 | 254 | ||||
GenBank mRNA | BC008048.2 | - | 1 | 254 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI711783.1 | - | 1 | 31 | Chr. 2 - 222318701 - 222342541 [+] on Build GRCh38 |
