siRNA Details
Gene Symbol: LOC101929715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110597.1 | 2 | 175 | |||
GenBank mRNA | BC111372.1 | - | 1 | 422 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR621233.1 | - | 1 | 1349 | Chr. 2 - 10448735 - 10450705 [+] on Build GRCh38 |
