siRNA Details
Gene Symbol: PRKG1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC039504.2 | - | 3, 4 | 241 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM479024.1 | - | 3, 4 | 254 | Chr. 10 - 52252147 - 52297917 [-] on Build GRCh38 |
