siRNA Details
Gene Symbol: LOC101927606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131977.1 | 2 | 452 | |||
GenBank mRNA | BC039507.1 | - | 2 | 451 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM478646.1 | - | 450 | Chr. - - [-] on Build |
