siRNA Details
Gene Symbol: LOC101927053 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110209.1 | 3 | 596 | |||
GenBank mRNA | BC068584.1 | - | 3 | 596 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB996525.1 | - | 3 | 623 | Chr. 2 - 96919106 - 96924365 [+] on Build GRCh38 |
