siRNA Details
Gene Symbol: ZNF391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076781.2 | NP_001070249.1 | 3 | 3304 | ||
NM_001322288.1 | NP_001309217.1 | 4 | 3528 | |||
NM_001322289.1 | NP_001309218.1 | 4 | 3383 | |||
NM_001322293.1 | NP_001309222.1 | 3 | 3272 | |||
XM_005249082.3 | XP_005249139.1 | 3 | 3089 | |||
XM_006715086.2 | XP_006715149.1 | 3 | 3030 | |||
XM_011514569.1 | XP_011512871.1 | 3 | 3012 | |||
GenBank mRNA | BC035154.2 | - | 1 | 1247 |
