siRNA Details
Gene Symbol: LOC100507388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047744.1 | - | 1 | 409 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ956614.1 | - | 1 | 408 | Chr. 4 - 74965281 - 74970123 [-] on Build GRCh38 |
