siRNA Details
Gene Symbol: LOC101927531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040862.2 | - | 2 | 233 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM800740.1 | - | 2 | 244 | Chr. 12 - 126359524 - 126360884 [+] on Build GRCh38 |
