siRNA Details
Gene Symbol: ABHD17C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091780.1 | - | 1 | 85 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK096008.1 | - | 1 | 689 | Chr. 7 - 132456130 - 132457937 [-] on Build GRCh38 |
