siRNA Details
Gene Symbol: LINC01016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038989.1 | 3 | 472 | |||
GenBank mRNA | AK057709.1 | - | 3 | 470 | ||
BC034285.1 | - | 1 | 532 | |||
BC042121.1 | - | 2 | 305 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB247614.1 | - | 3 | 470 | Chr. 6 - 33892103 - 33896907 [-] on Build GRCh38 |
