siRNA Details
Gene Symbol: LOC101927374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110133.1 | 3 | 1698 | |||
GenBank mRNA | AK094694.1 | - | 3 | 1698 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110133.1 | 3 | 1698 | |||
GenBank mRNA | AK094694.1 | - | 3 | 1698 |