siRNA Details
Gene Symbol: LINC01153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037739.1 | - | 2 | 479 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG724023.1 | - | 2 | 479 | Chr. 10 - 121178700 - 121183674 [+] on Build GRCh38 |
