siRNA Details
Gene Symbol: LOC101928446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110938.1 | 2 | 3376 | |||
GenBank mRNA | AL833113.1 | - | 2 | 3379 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110938.1 | 2 | 3376 | |||
GenBank mRNA | AL833113.1 | - | 2 | 3379 |