siRNA Details
Gene Symbol: TCONS_00029157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131902.1 | 1 | 191 | |||
GenBank mRNA | AL530227.3 | - | 1 | 191 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR609588.1 | - | 1 | 191 | Chr. 21 - 43358184 - 43362349 [-] on Build GRCh38 |
