siRNA Details
Gene Symbol: LINC00315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF427488.1 | - | 3 | 2097 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BP378314.1 | - | 1 | 200 | Chr. 21 - 45300102 - 45300669 [-] on Build GRCh38 |
