siRNA Details
Gene Symbol: LOC283745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097986.1 | - | 1 | 606 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CV572248.1 | - | 1 | 506 | Chr. 15 - 40527773 - 40528403 [-] on Build GRCh38 |
