siRNA Details
Gene Symbol: ATP6V1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127836.1 | - | 1 | 545 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF370403.1 | - | 1 | 561 | Chr. 2 - 10776985 - 10779707 [+] on Build GRCh38 |
