siRNA Details
Gene Symbol: LOC100505915 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125434.1 | 1 | 196 | |||
GenBank mRNA | AK125313.1 | - | 1 | 196 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA719594.1 | - | 1 | 196 | Chr. 16 - 15094411 - 15103313 [+] on Build GRCh38 |
