siRNA Details
Gene Symbol: ANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008067.1 | XP_016863556.1 | 1 | 960 | ||
XM_017008069.1 | XP_016863558.1 | 1 | 959 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AB209779.1 | - | 1370 | Chr. - - [-] on Build |
