siRNA Details
Gene Symbol: LOC158434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132344.1 | 3 | 420 | |||
GenBank mRNA | AK055173.1 | - | 3 | 421 | ||
AL041156.1 | - | 1 | 389 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA434413.1 | - | 3 | 421 | Chr. 9 - 96108804 - 96116411 [-] on Build GRCh38 |
