siRNA Details
Gene Symbol: LOC158434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132344.1 | 3 | 1476 | |||
GenBank mRNA | AK055173.1 | - | 3 | 1477 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC158434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132344.1 | 3 | 1476 | |||
GenBank mRNA | AK055173.1 | - | 3 | 1477 |