siRNA Details
Gene Symbol: NCOA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012970.1 | XP_016868459.1 | 1 | 4768 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK025045.1 | - | 1 | 324 | Chr. 8 - 70218200 - 70220303 [-] on Build GRCh38 |
