siRNA Details
Gene Symbol: DYNC2LI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040558.1 | - | 6 | 573 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF242772.1 | - | 1 | 870 | Chr. 1 - 50954541 - 50956665 [-] on Build GRCh38 |
