siRNA Details
Gene Symbol: C14orf159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286470.1 | NP_001273399.1 | 3, 4 | 698 | ||
GenBank mRNA | AK123532.1 | - | 3, 4 | 341 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK123973.1 | - | 2 | 1722 | Chr. 14 - 91146674 - 91149247 [+] on Build GRCh38 |
