siRNA Details
Gene Symbol: ZNF862 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054924.1 | - | 1 | 1411 | ||
BC045757.1 | - | 1 | 1406 | |||
BC092506.1 | - | 1 | 1436 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF862 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054924.1 | - | 1 | 1411 | ||
BC045757.1 | - | 1 | 1406 | |||
BC092506.1 | - | 1 | 1436 |