siRNA Details
Gene Symbol: LOC401134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033976.1 | 7 | 1778 | |||
GenBank mRNA | BC040544.2 | - | 7 | 1779 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC401134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033976.1 | 7 | 1778 | |||
GenBank mRNA | BC040544.2 | - | 7 | 1779 |