siRNA Details
Gene Symbol: LOC101928539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109900.1 | 1 | 1966 | |||
GenBank mRNA | BC041836.1 | - | 1 | 1966 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109900.1 | 1 | 1966 | |||
GenBank mRNA | BC041836.1 | - | 1 | 1966 |