siRNA Details
Gene Symbol: RIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177663.1 | NP_001171134.1 | 35 | 9134 | ||
NM_001177664.1 | NP_001171135.1 | 35 | 9037 | |||
NM_001177665.1 | NP_001171136.1 | 34 | 9307 | |||
NM_018151.4 | NP_060621.3 | 36 | 9212 | |||
XM_005246665.3 | XP_005246722.1 | 36 | 9092 | |||
XM_011511395.2 | XP_011509697.1 | 35 | 6348 | |||
XM_017004422.1 | XP_016859911.1 | 36 | 9161 | |||
XM_017004423.1 | XP_016859912.1 | 35 | 8999 | |||
XM_017004424.1 | XP_016859913.1 | 35 | 9029 | |||
XM_017004425.1 | XP_016859914.1 | 34 | 9337 | |||
GenBank mRNA | BC072675.1 | - | 1 | 578 |
