siRNA Details
Gene Symbol: ANKRD44-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046837.1 | 2 | 323 | |||
GenBank mRNA | BC042827.1 | - | 2 | 323 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI463461.1 | - | 2 | 327 | Chr. 2 - 197299485 - 197302521 [-] on Build GRCh38 |
