siRNA Details
Gene Symbol: FAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000043.5 | NP_000034.1 | 9 | 2907 | ||
NM_001320619.1 | NP_001307548.1 | 8 | 2824 | |||
NM_152871.3 | NP_690610.1 | 8 | 2844 | |||
NM_152872.3 | NP_690611.1 | 8 | 2882 | |||
NR_028033.3 | 7 | 2735 | ||||
NR_028034.3 | 6 | 2597 | ||||
NR_028035.3 | 7 | 2660 | ||||
NR_028036.3 | 8 | 2798 | ||||
NR_135313.1 | 7 | 2715 | ||||
NR_135314.1 | 9 | 2898 | ||||
NR_135315.1 | 7 | 2651 | ||||
GenBank mRNA | AK026195.1 | - | 1 | 196 |
