siRNA Details
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257158.1 | NP_001244087.1 | 10 | 3835 | ||
NM_001257159.1 | NP_001244088.1 | 9 | 3787 | |||
NM_018718.2 | NP_061188.1 | 11 | 4051 | |||
NR_046443.1 | 11 | 3975 | ||||
XM_011516709.2 | XP_011515011.1 | 12 | 4012 | |||
XM_011516710.2 | XP_011515012.1 | 10 | 3827 | |||
GenBank mRNA | BC047500.1 | - | 1 | 1549 |
