siRNA Details
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257158.1 | NP_001244087.1 | 10 | 4465 | ||
NM_001257159.1 | NP_001244088.1 | 9 | 4417 | |||
NM_018718.2 | NP_061188.1 | 11 | 4681 | |||
NR_046443.1 | 11 | 4605 | ||||
GenBank mRNA | BC047500.1 | - | 1 | 2182 |
