siRNA Details
Gene Symbol: TTC21B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038983.1 | 5 | 606 | |||
NR_038984.1 | 4 | 550 | ||||
GenBank mRNA | AK123487.1 | - | 4 | 550 | ||
DA373445.1 | - | 4 | 550 | |||
DA374147.1 | - | 4 | 550 | |||
DA378622.1 | - | 4 | 414 |
