siRNA Details
Gene Symbol: TTC21B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038983.1 | 5 | 635 | |||
NR_038984.1 | 4 | 579 | ||||
GenBank mRNA | AK123487.1 | - | 4 | 579 | ||
DA374147.1 | - | 4 | 579 | |||
DA378622.1 | - | 4 | 443 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA383109.1 | - | 4 | 445 | Chr. 2 - 165933991 - 165948318 [+] on Build GRCh38 |
