siRNA Details
Gene Symbol: TTC21B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038984.1 | 1, 2 | 212 | |||
GenBank mRNA | AK123487.1 | - | 1, 2 | 212 | ||
DA373445.1 | - | 1, 2 | 212 | |||
DA374147.1 | - | 1, 2 | 212 | |||
DA378622.1 | - | 1, 2 | 76 |
