siRNA Details
Gene Symbol: LINC00485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033855.1 | 1, 2 | 100 | |||
GenBank mRNA | BC037211.1 | - | 1, 2 | 100 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG772150.1 | - | 1, 2 | 105 | Chr. 12 - 102812221 - 102824399 [-] on Build GRCh38 |
