siRNA Details
Gene Symbol: LOC101928336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110396.1 | 2, 3 | 219 | |||
GenBank mRNA | BC041420.1 | - | 2, 3 | 219 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE778080.1 | - | 198 | Chr. - - [-] on Build |
