siRNA Details
Gene Symbol: LINC00152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC070202.1 | - | 106 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD639006.1 | - | 1, 2 | 109 | Chr. 2 - 87509214 - 87521518 [+] on Build GRCh38 |
