siRNA Details
Gene Symbol: LOC105372392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123379.1 | - | 2 | 2033 |
Gene Symbol: LINC01535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110720.1 | 4 | 516 | |||
GenBank mRNA | BX099288.1 | - | 4 | 478 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX089579.1 | - | 6 | 411 | Chr. 19 - 36316039 - 36331710 [-] on Build GRCh38 |
